MilliporeSigma
  • Synthesis of dimeric quinazolin-2-one, 1,4-benzodiazepin-2-one, and isoalloxazine compounds as inhibitors of amyloid peptides association.
Synthesis of dimeric quinazolin-2-one, 1,4-benzodiazepin-2-one, and isoalloxazine compounds as inhibitors of amyloid peptides association.
Archiv der Pharmazie (2009-07-01)
Alexander Barthel, Lothar Trieschmann, Dieter Ströhl, Ralph Kluge, Gerald Böhm, René Csuk
ABSTRACT
The synthesis of dimeric compounds derived from quinazolin-2-one and 1,4-benzodiazepin-2-one possessing a piperazine or homopiperazine spacer was investigated. In addition, a piperazine spacered bis-isoalloxazine and a bis-riboflavin compound were prepared and their ability to interrupt the association of prion proteins and Alzheimer-specific Abeta peptides was investigated using a fast screening system based on flow cytometry. The bis-isoalloxazine 14 was identified as a new lead structure.
Sigma-Aldrich
Alloxazine, 96%