MilliporeSigma
[Studies on chemical constituents of herbs of Oxytropis microphylla].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2009-07-31)
Cheng Peng, Zhongduo Yang
ABSTRACT
To study the chemical constituents of Oxytropis microphylla. The compounds were isolated and purified by various column chromatographic techniques and their structures were elucidated on the basis of spectral analysis (EI-MS, 1H-NMR, 13C-NMR). Eight compounds were isolated and identified as kaempferol (1), baicalein (2), 5-hydroxy-7, 8, 4'-trimethoxyflavone (3), stigmasterol (4), stigmast-4-en-3beta-ol (5), beta-sitosterol (6), lupenol (7), dehydrocostus lactone (8). All compounds were isolated for the first time from this plant, compounds 2, 3, 5, and 8 were isolated for the first time from this genus.