MilliporeSigma
  • The efficacy of three class I topical synthetic corticosteroids, fluocinonide 0.1% cream, clobetasol 0.05% cream and halobetasol 0.05% cream: a Scholtz-Dumas bioassay comparison.
The efficacy of three class I topical synthetic corticosteroids, fluocinonide 0.1% cream, clobetasol 0.05% cream and halobetasol 0.05% cream: a Scholtz-Dumas bioassay comparison.
Journal of drugs in dermatology : JDD (2009-08-12)
Chai Sue Lee, John Koo
ABSTRACT
This study compared the efficacy of a novel, topical class I synthetic, 0.10% fluocinonide corticosteroid with two other class I corticosteroids and placebo for the treatment of plaque psoriasis. A 0.5 gram dose of fluocinonide 0.1% cream, clobetasol propionate 0.05% cream, halobetasol propionate 0.05% cream, and placebo ointment were applied to test sites on one psoriatic plaque per patient (n=5). Test sites were outlined according to the Scholtz-Dumas bioassay. Test sites were assessed by a blinded evaluator (1 = psoriasis worsened to 5 = psoriasis clear or almost clear), cleaned and medications were reapplied on days 3, 5, 7, 10 and 12. The three class I corticosteroid products were comparably effective, numerically and statistically, in clearing the psoriatic plaques. Upon completion of treatment, 60-80% of active-treated sites were clear or almost clear of psoriasis compared to zero with the placebo.
Sigma-Aldrich
Fluocinonide, ≥98% (HPLC)