MilliporeSigma
  • [Protective effects of astilbin on renal ischemia-reperfusion injury in rats].
[Protective effects of astilbin on renal ischemia-reperfusion injury in rats].
Zhong xi yi jie he xue bao = Journal of Chinese integrative medicine (2009-08-13)
Shao-Hua Song, Xiao-Yun Shen, Fang Liu, Yi Tang, Zhen-Meng Wang, Zhi-Ren Fu
ABSTRACT
To investigate the protective effects of astilbin on renal ischemia-reperfusion (IR) injury in rats. Twenty-four male SD rats, two months old, were randomly allocated into three groups: sham-operated group (n=8), untreated group (n=8) and astilbin group (n=8). Rats in the untreated group and the astilbin group underwent temporary renal artery occlusion to induce IR injury. The rats in the astilbin group were intraperitoneally injected with 12 mg/mL astilbin at a dose of 30 mg/kg from 3 day before IR injury until to be sacrificed once per day, and rats in the untreated group were injected with equal volume of normal saline at the same time. After 6-hour reperfusion, blood urea nitrogen (BUN) and serum creatinine (SCr) and histological changes of the renal tissues were detected to evaluate renal injury. Expressions of monocyte chemoattractant protein-1 (MCP-1) mRNA and protein in the renal tissues and the serum contents of interleukin-6 (IL-6) and IL-1beta were also measured with semi-quantitative reverse transcription-polymerase chain reaction, Western blotting or enzyme-linked immunosorbent assay. Compared with the untreated group, BUN and SCr levels were significantly decreased in the astilbin group after 6-hour reperfusion (P<0.01), and similar results were also found in histological examination. The expressions of MCP-1 mRNA and protein in renal tissues in the astilbin group were lower than those in the untreated group. The serum contents of IL-6 and IL-1beta were decreased in the astilbin group as compared with the untreated group (P<0.01). Astilbin can ameliorate kidney IR injury in rats by inhibiting the production of chemokine MCP-1 and cytokines IL-6 and IL-1beta.
Sigma-Aldrich
Astilbin from Engelhardtia roxburghiana, ≥98%