MilliporeSigma
  • Excited state intramolecular proton transfer in 3-hydroxy flavone and 5-hydroxy flavone: a DFT based comparative study.
Excited state intramolecular proton transfer in 3-hydroxy flavone and 5-hydroxy flavone: a DFT based comparative study.
Journal of molecular modeling (2009-09-18)
Sankarlal Ash, Sankar Prasad De, Santanu Pyne, Ajay Misra
ABSTRACT
Potential energy (PE) curves for the intramolecular proton transfer in the ground (GSIPT) and excited (ESIPT) states of 3-hydroxy-flavone (3HF) and 5-hydroxy-flavone (5HF) were studied using DFT/B3LYP (6-31G (d,p)) and TD-DFT/B3LYP (6-31G (d,p)) level of theory respectively. Our calculations suggest the non-viability of ground state intramolecular proton transfer for both the compounds. Calculated PE curves of 3HF for the ground and excited singlet states proton transfer process explain its four state laser diagram. Excited states PE calculations support the ESIPT process to both 5HF and 3HF. The difference in ESIPT emission process of 3HF and 5HF have been explained in terms of HOMO and LUMO electron distribution of the enol and keto tautomer of these two compounds.