MilliporeSigma
  • [Derivatives of para-hydroxyacetophenone and para-hydroxypropiophenone as a potential new beta-adrenolytic agent].
[Derivatives of para-hydroxyacetophenone and para-hydroxypropiophenone as a potential new beta-adrenolytic agent].
Ceskoslovenska farmacie (1990-11-01)
R Cizmáriková, A Dingová, J Kozlovský, O Greksáková
ABSTRACT
Within the framework of the study of the structure-effect relationship, a series of novel potential beta-adrenolytic agents derived from p-hydroxyacetophenone and p-hydroxypropiophenone with an alyloxymethyl and a cycloalkyloxymethyl group in the lipophilic part of the molecule and an isopropyl and a tert-butyl group in the hydrophilic part on the basic nitrogen were prepared by means of a well-tried method. The structure of the prepared drugs was confirmed on the basis of interpretation of the IR, UV, 1H-NMR and mass spectra. The results of pharmacological evaluation of selected drugs show that the agents poses beta-adrenolytic and antiarrhythmic activity. From the viewpoint of comparison of the individual drugs subjected to testing the presence of an alyl seems more advantageous than that of a cyclohexyl. The characteristic of the prepared drugs was supplemented by the determination of their partition coefficients, surface tension and acute toxicity.