MilliporeSigma
  • Synthesis of (E,Z)-1-alkoxy-3-acyloxy-2-methylpenta-1,3-dienes via Danishefsky-type dienes or O-acylation of enones.
Synthesis of (E,Z)-1-alkoxy-3-acyloxy-2-methylpenta-1,3-dienes via Danishefsky-type dienes or O-acylation of enones.
The Journal of organic chemistry (2009-10-22)
Sylvain Laclef, Claudia J Exner, Maris Turks, Valeria Videtta, Pierre Vogel
ABSTRACT
1-Alkoxy-2-methyl-3-acyloxy-(E,E)-penta-1,3-dienes have been prepared applying among others a modified Danishefsky's general method, including chiral, racemic, and achiral derivatives.