MilliporeSigma
  • A combined theoretical and experimental investigation on the enantioselective oxidation of aryl benzyl sulfides in the presence of a chiral titanium catalyst.
A combined theoretical and experimental investigation on the enantioselective oxidation of aryl benzyl sulfides in the presence of a chiral titanium catalyst.
Chemistry (Weinheim an der Bergstrasse, Germany) (2009-11-13)
Francesco Naso, Maria Annunziata M Capozzi, Andrea Bottoni, Matteo Calvaresi, Valerio Bertolasi, Francesco Capitelli, Cosimo Cardellicchio
ABSTRACT
An experimental investigation of the enantioselective oxidation of aryl benzyl sulfides by tert-butyl hydroperoxide in the presence of a titanium/hydrobenzoin catalyst has shown that these sulfides are ideal substrates for this catalytic system, with negligible interference by the substituents on the aryl groups. A reaction mechanism based on DFT computations has been proposed. The DFT MPWB1K functional was used in the theoretical investigation to account for weak hydrogen-bonding and pi interactions. The computed reaction profile explains the experimentally observed enantioselectivity, which is determined by the thermodynamics of the first phase of the reaction. A detailed discussion of the hydrogen-bonding and pi interactions that drive the reaction along the observed stereochemical path is given.
Sigma-Aldrich
Dibenzyl sulfide, ≥95.0% (HPLC)