MilliporeSigma
  • DNA interstrand cross-linking activity of (1-Chloroethenyl)oxirane, a metabolite of beta-chloroprene.
DNA interstrand cross-linking activity of (1-Chloroethenyl)oxirane, a metabolite of beta-chloroprene.
Chemical research in toxicology (2009-12-25)
Brian A Wadugu, Christopher Ng, Bethany L Bartley, Rebecca J Rowe, Julie T Millard
ABSTRACT
With the goal of elucidating the molecular and cellular mechanisms of chloroprene toxicity, we examined the potential DNA cross-linking of the bifunctional chloroprene metabolite, (1-chloroethenyl)oxirane (CEO). We used denaturing polyacrylamide gel electrophoresis to monitor the possible formation of interstrand cross-links by CEO within synthetic DNA duplexes. Our data suggest interstrand cross-linking at deoxyguanosine residues within 5'-GC and 5'-GGC sites, with the rate of cross-linking depending on pH (pH 5.0 > pH 6.0 > pH 7.0). A comparison of the cross-linking efficiencies of CEO and the structurally similar cross-linkers diepoxybutane (DEB) and epichlorohydrin (ECH) revealed that DEB > CEO > or = ECH. Furthermore, we found that cytotoxicity correlates with cross-linking efficiency, supporting a role for interstrand cross-links in the genotoxicology of chloroprene.