MilliporeSigma
  • The hydrated and anhydrous gold(III) tetrachloride salts of L-ecgonine, an important forensic toxicology marker for cocaine.
The hydrated and anhydrous gold(III) tetrachloride salts of L-ecgonine, an important forensic toxicology marker for cocaine.
Acta crystallographica. Section C, Crystal structure communications (2010-01-06)
Matthew R Wood, Hugh W Thompson, Thomas A Brettell, Roger A Lalancette
ABSTRACT
The structure of the hydrated gold(III) tetrachloride salt of L-ecgonine {hydronium tetrakis[(1R,2R,3S,5S,8S)-3-hydroxy-8-methyl-8-azoniabicyclo[3.2.1]octane-2-carboxylate pentakis[tetrachloridoaurate(III)] hexahydrate}, (C(9)H(16)NO(3))(4)(H(3)O)[AuCl(4)](5).6H(2)O, demonstrates an unprecedented stoichiometric relationship between the cations and anions in the unit cell. The previous tropane alkaloid structures, including the related hydrochloride salts, all have a cation-anion ratio of 1:1, as does the anhydrous salt described here, namely (1R,2R,3S,5S,8S)-3-hydroxy-8-methyl-8-azoniabicyclo[3.2.1]octane-2-carboxylate tetrachloridoaurate(III), (C(9)H(16)NO(3))[AuCl(4)]. The hydrated salt, however, consists of four monopositive N-protonated units of the alkaloid and five [AuCl(4)](-) counter-ions, plus seven solvent water molecules. The H atom required for change balance has been assigned to a water molecule. In addition, the hydrate has a novel arrangement, with all seven of the water molecules and all of the O atoms in the cations participating in an alternating arrangement of interleaved sheets of the anionic species. Both the hydrate and the anhydrous salt of the same toxicologically important marker for cocaine show that the cation and anion are in close proximity to each other, as was found in the gold(III) tetrachloride salt of L-cocaine.