MilliporeSigma
  • 19-hydroxyandrostenedione does not modulate [3H]aldosterone binding to human mononuclear leucocytes and rat renal cytosol.
19-hydroxyandrostenedione does not modulate [3H]aldosterone binding to human mononuclear leucocytes and rat renal cytosol.
The Journal of steroid biochemistry and molecular biology (1991-03-01)
H Murase, K Yasuda, L B Mercado-Asis, A Mori, T Shimada, T Mune, H Morita, N Noritake, N Yamakita, K Miura
ABSTRACT
To verify the aldosterone amplifying action of 19-hydroxyandrostenedione (19-OH-AD), we investigated [3H]aldosterone and [3H]19-OH-AD binding to type I (mineralocorticoid) receptor in the renal cytosol of adrenalectomized and ovariectomized rat, and human mononuclear leucocytes (MNL). In the [3H]aldosterone binding study, the cytosol was incubated with [3H]aldosterone and 200-fold RU28362 (11 beta,17 beta-dihydroxy-6-methyl,17 alpha-(1-propynyl)-androsta-1,4,6- trien-3-one), a pure glucocorticoid, with or without 19-OH-AD. Scatchard plots of [3H]aldosterone binding to cytosol with 0.2 or 20 nM 19-OH-AD or without 19-OH-AD were linear. Dissociation constants (Kd) and maximum bindings (Bmax) without 19-OH-AD, and with 0.2 and 20 nM 19-OH-AD were: 0.71 +/- 0.03 nM and 23.0 +/- 3.4 fmol/mg protein (mean +/- SD, n = 3), 0.72 +/- 0.05 nM and 23.1 +/- 2.3 fmol/mg protein (n = 3), and 0.77 +/- 0.04 nM and 22.9 +/- 4.8 fmol/mg protein (n = 3), respectively. 19-OH-AD did not significantly change the Kd and Bmax of [3H]aldosterone binding. A high concentration of 19-OH-AD slightly displaced 0.2 or 5 nM [3H]aldosterone bound to cytosol. In human MNL, Scatchard plots of [3H]aldosterone binding with both 0.2 and 20 nM 19-OH-AD and without 19-OH-AD were linear. Kd and Bmax were, respectively, 1.00 nM and 780 sites/cell in the absence of 19-OH-AD, and 1.07 nM and 774 sites/cell in the presence of 0.2 nM 19-OH-AD. Without 19-OH-AD they were, respectively, 0.95 nM and 551 sites/cell, and 1.10 nM and 560 sites/cell with 20 nM 19-OH-AD. A high concentration of 19-OH-AD slightly displaced 0.2 or 5 nM of [3H]aldosterone bound to MNL. In both tissues, there was no obvious specific binding of [3H]19-OH-AD within the range of 1-60 nM. The above results suggest that the amplifying effect of 19-OH-AD on aldosterone mineralocorticoid action may not occur at the binding site of aldosterone to type I receptor, and that 19-OH-AD itself may not have any direct or indirect mineralocorticoid actions on the steroid receptor-mediated process in the rat kidney and human MNL.