MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Current development and patents on high-brightness white LED for illumination.
Current development and patents on high-brightness white LED for illumination.
Recent patents on nanotechnology (2010-03-11)
Wen-Yuan Pang, Ikai Lo, Chia-Ho Hsieh, Yu-Chi Hsu, Ming-Chi Chou, Cheng-Hung Shih
ABSTRACT
In this paper, we reviewed the current development and patents for the application of high-brightness and high-efficiency white light-emitting diode (LED). The high-efficiency GaN nanostructures, such as disk, pyramid, and rod were grown on LiAlO(2) substrate by plasma-assisted molecular-beam epitaxy, and a model was developed to demonstrate the growth of the GaN nanostructures. Based on the results, the GaN disk p-n junction was designed for the application of high brightness and high efficiency white LED.
Sigma-Aldrich
Lithium aluminate