MilliporeSigma
  • Evidence for unique and ubiquitous environmental sources of 3,3'-dichlorobiphenyl (PCB 11).
Evidence for unique and ubiquitous environmental sources of 3,3'-dichlorobiphenyl (PCB 11).
Environmental science & technology (2010-04-14)
Lisa A Rodenburg, Jia Guo, Songyan Du, Gregory J Cavallo
ABSTRACT
The non-Aroclor congener 3,3'-dichlorobiphenyl (PCB 11) has been recently detected in air, water, biota, sediment, and suspended sediment. Although it has been known since at least the 1970s that this congener is produced inadvertently during the production of diarylide yellow pigments, this work presents the first evidence that the use of these pigments in consumer goods results in the dispersion of PCB 11 throughout the environment at levels that are problematic in terms of achieving water quality standards for the sum of polychlorinated biphenyls (PCBs). In this work, PCB 11 is measured at ppb levels in consumer goods that are likely to be discarded in ways that allow them to enter wastewater treatment plants and combined sewer overflows, including newspapers, magazines, cardboard boxes used for food packaging, and plastic bags. Also, using data sets acquired for the purpose of calculating total maximum daily loads (TMDLs) for PCBs, PCB 11 loads to the New York/New Jersey Harbor and Delaware River are calculated. Despite the fact that there are no known manufacturers of diarylide yellow pigments in the Delaware River watershed, the loads of PCB 11 to the Delaware River exceed the TMDL for the sum of PCBs by nearly a factor of 2. The ratio of PCB 11 to a characteristic dechlorination end product, PCB 4 (2,2'-dichlorobiphenyl), in these data sets indicates that dechlorination is not a significant source of PCB 11 in these systems. In the upper Hudson River, where extensive dechlorination of heavy PCB congeners occurs, the ratio is just 0.012. In contrast, downstream in the NY/NJ Harbor as well as in the Delaware River the ratio is much higher and more variable. Pigment use therefore appears to be the main source of PCB 11 in these systems, and this congener is likely to present a significant obstacle to achieving PCB water quality standards throughout the United States.