MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Agmatine, an endogenous imidazoline receptor ligand modulates ethanol anxiolysis and withdrawal anxiety in rats.
Agmatine, an endogenous imidazoline receptor ligand modulates ethanol anxiolysis and withdrawal anxiety in rats.
European journal of pharmacology (2010-04-17)
Brijesh G Taksande, Nandkishor R Kotagale, Mital R Patel, Gajanan P Shelkar, Rajesh R Ugale, Chandrabhan T Chopde
ABSTRACT
Present study investigated the role of agmatine in ethanol-induced anxiolysis and withdrawal anxiety using elevated plus maze (EPM) test in rats. The anxiolytic-like effect of ethanol was potentiated by pretreatment with imidazoline I(1)/I(2) receptor agonist agmatine (10-20 mg/kg, i.p.), imidazoline I(1) receptor agonists, moxonidine (0.25 mg/kg, i.p.) and clonidine (0.015 mg/kg, i.p.), imidazoline I(2) receptor agonist, 2-BFI (5 mg/kg, i.p.) as well as by the drugs known to increase endogenous agmatine levels in brain viz., L-arginine, an agmatine biosynthetic precursor (100 microg/rat, i.c.v.), ornithine decarboxylase inhibitor, DFMO (125 microg/rat, i.c.v.), diamine oxidase inhibitor, aminoguanidine (65 microg/rat, i.c.v.) and agmatinase inhibitor, arcaine (50 microg/rat, i.c.v.). Conversely, prior administration of I(1) receptor antagonist, efaroxan (1 mg/kg, i.p.), I(2) receptor antagonist, idazoxan (0.25mg/kg, i.p.) and arginine decarboxylase inhibitor, D-arginine (100 microg/rat, i.c.v.) blocked the anxiolytic-like effect of ethanol. Moreover, ethanol withdrawal anxiety was markedly attenuated by agmatine (10-20 mg/kg, i.p.), moxonidine (0.25 mg/kg, i.p.), clonidine (0.015 mg/kg, i.p.), 2-BFI (5 mg/kg, i.p.), L-arginine (100 microg/rat, i.c.v.), DFMO (125 microg/rat, i.c.v.), aminoguanidine (65 microg/rat, i.c.v.) and arcaine (50 microg/rat, i.c.v.). The anti-anxiety effect of agmatine in ethanol-withdrawn rats was completely blocked by efaroxan (1 mg/kg, i.p.) and idazoxan (0.25 mg/kg, i.p.). These results suggest that agmatine and imidazoline receptor system may be implicated in ethanol-induced anxiolysis and withdrawal anxiety and strongly support further investigation of agmatine in ethanol dependence mechanism. The data also project agmatine as a potential therapeutic target in overcoming alcohol withdrawal symptoms such as anxiety.
Sigma-Aldrich
Arcaine sulfate salt