MilliporeSigma
[Study on quality standards for Lingmaoxiang Jiedu pills].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2010-04-29)
Cheng Zheng, Binbin Zheng, Tongwei Yao
ABSTRACT
To establish the quality standards of Linmaoxiang Jiedu pills. Cinobufagin and bufogenin were determined by HPLC simultaneously. The average recoverys of Cinobufagin and bufogenin was 100.5% and 100.3%, their linear range were 48.74-731.10 ng and 49.90-748.50 ng, respectively. The method for quantification is reproducible and realizable. The method can be used to control quality of Linmaoxiang Jiedu pills as the quality standards.