MilliporeSigma
[Chemical constituents from Sarcandra glabra].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2010-06-16)
Chao Wang, Liping Zhu, Jingzhi Yang, Chuangjun Li, Dongming Zhang
ABSTRACT
To study the chemical constituents of the plant of Sarcandra glabra and provide reference for the study of the bioactive substances. The compounds were isolated from the EtOH extract by various chromatographic methods and their structures were elucidated by their physico-chemical properties and the analysis of their spectral data. Nine compounds were isolated and identified as isoscopletin (1), syringaresinol monoside (2), styraxjaponoside B (3), 5-O-caffeoylshikimic acid (4), shizukanolide E (5), isoastilbin (6), neoisoastilbin (7), astilbin (8), neoastilbin (9). Compounds 1-7 were isolated from S. glabra for the first time.