MilliporeSigma
  • Ca2+ dependence of motilide-induced contractions in rabbit duodenal muscle strips in vitro.
Ca2+ dependence of motilide-induced contractions in rabbit duodenal muscle strips in vitro.
Naunyn-Schmiedeberg's archives of pharmacology (1991-02-01)
T L Peeters, G Matthijs, G Vantrappen
ABSTRACT
Recent studies suggested that certain erythromycin A (EM-A) derivatives are motilin receptor agonists. As proposed by Itoh they may be called "motilides". We have investigated the Ca2(+)-dependence of contractions induced by two potent motilides, ME-34 [de(N-methyl) 8,9-anhydroeryhtromycin A 6,9-hemiacetal] and EM-523 [de(N-methyl)-N-ethyl-8,9-anhydroerythromycin A 6,9-hemiacetal], in duodenal tissues and compared the results with those previously obtained with motilin. Isometric and isotonic contractile responses of isolated longitudinal muscle sheets from the rabbit duodenum were tested under normal, Ca2(+)-free and depolarizing conditions. Prior to stimulation with motilides, the maximal response to acetylcholine was recorded and all responses were always expressed as a percentage of this response. Both motilides induced contractions in normally polarized tissue, with an EC50 of 26 +/- 5 nM for ME-34 (n = 7), and 27 +/- 5 nM for EM-523 (n = 16) and maximal responses of respectively 88 +/- 4% and 80 +/- 3%. Like motilin, both compounds induced an 'extra'-contraction in depolarized tissues. The EM-523 response in 140 mM K+ under isotonic conditions was 84 +/- 3% (n = 5) at 10(-5) M, with an EC50 that was shifted to 65 +/- 18 nM. Similar figures were obtained for ME-34. When Ca2+ was added to Ca2(+)-depleted strips, half-maximal Ca2+ values (in mM) were 1.10 +/- 0.11 (n = 9) for EM-523 and 1.13 +/- 0.12 (n = 3) for ME-34, as compared with 1.12 +/- 0.13 (n = 7) for motilin and 2.8 +/- 1.1 (n = 9) for K+. Both ME-34 and EM-523 also induced a transient contraction in Ca2(+)-free solutions under isometric conditions.(ABSTRACT TRUNCATED AT 250 WORDS)
Product Description
Supelco
Erythromycin A enol ether, analytical standard