MilliporeSigma
  • In vitro approaches to evaluate placental drug transport by using differentiating JEG-3 human choriocarcinoma cells.
In vitro approaches to evaluate placental drug transport by using differentiating JEG-3 human choriocarcinoma cells.
Basic & clinical pharmacology & toxicology (2010-10-20)
Kenji Ikeda, Naoki Utoguchi, Hidenobu Tsutsui, Satoko Yamaue, Manami Homemoto, Erina Nakao, Yumi Hukunaga, Kyohei Yamasaki, Michiaki Myotoku, Yoshihiko Hirotani
ABSTRACT
Human choriocarcinoma cells have been used as models for studying transcellular drug transport through placental trophoblasts. However, these models allow the transport of low-molecular-weight drugs through intercellular gap junctions. This study aimed at investigating the differentiation patterns of JEG-3 choriocarcinoma cells under different culture conditions and establishing the appropriate model of in vitro syncytiotrophoblast drug transport. Paracellular permeability was estimated by measuring the transepithelial electrical resistance (TEER) across JEG-3 cell layers. The mRNA expression levels of non-expressed in choriocarcinoma clone 1 (NECC1) and breast cancer resistance protein (BCRP), and those of E-cadherin (ECAD) and cadherin-11 (CDH11), which are adherens junction-associated proteins related to fusogenic ability of syncytiotrophoblasts differentiated from cytotrophoblasts, protein expression levels were considered as the differentiation signals. The highest TEER values were obtained in the JEG-3 cells cultured in the Dulbecco's modified Eagle's medium (DMEM)/Ham's F-12 (1:1) mixed medium (CS-C(®) ; Dainippon Sumitomo Pharma Co. Ltd., Osaka, Japan). By comparing the TEER values and the differentiation signals, the authors identified at least five JEG-3 cell-differentiation patterns. The differentiation pattern of JEG-3 cultured in CS-C resembled the syncytiotrophoblast-like differentiation signal characterizations in vivo. In conclusion, the syncytiotrophoblast-like models of differentiating JEG-3 cells cultured in CS-C might be appropriate for evaluating drug transport across the placental trophoblast.