MilliporeSigma
  • AdoHcy hydrolase of Trichomonas vaginalis: studies of the effects of 5'-modified adenosine analogues and related 6-N-cyclopropyl derivatives.
AdoHcy hydrolase of Trichomonas vaginalis: studies of the effects of 5'-modified adenosine analogues and related 6-N-cyclopropyl derivatives.
Bioorganic & medicinal chemistry letters (2010-11-04)
Padraick J Dornbush, Guillermo Vazquez-Anaya, Ajit Shokar, Saoly Benson, Magdalena Rapp, Stanislaw F Wnuk, Lisa A Wrischnik, Kirkwood M Land
ABSTRACT
Trypanosoma brucei and Trichomonas vaginalis are both parasitic protozoans that are known to share many similar biochemical pathways. Aristeromycin, as well as 5'-iodovinyl and 5'-oxime analogues of adenosine, are potent inhibitors of AdoHcy hydrolase in T. brucei, an enzyme that catalyses the hydrolysis of AdoHcy to adenosine and L-homocysteine. To help determine the role of this enzyme in T. vaginalis, we have tested a library of 5'-modified adenosine derivatives, including 5'-deoxy-5'-(iodomethylene)-adenosine and related 6-N-cyclopropyl analogues. Our results indicate that these inhibitors are effective at inhibiting the growth of T. vaginalis, by as much as 95%.