MilliporeSigma
  • Synthesis and structure-activity relationships of fenbufen amide analogs.
Synthesis and structure-activity relationships of fenbufen amide analogs.
Molecules (Basel, Switzerland) (2010-12-04)
Kun-I Lin, Chao-Hsun Yang, Chia-Wen Huang, Jhen-Yi Jian, Yu-Chun Huang, Chung-Shan Yu
ABSTRACT
The previous discoveries of butyl fenbufen amide analogs with antitumor effects were further examined. The amide analogs with 1, 3, 4 and 8 carbons chains were prepared in 70-80% yield. Fenbufen had no cytotoxic effects at concentrations ranging from 10 to 100 μM. Methyl fenbufen amide had significant cytotoxic effects at a concentration of 100 μM. As the length of the alkyl amide side chain increased, the cytotoxic effects increased, and the octyl fenbufen amide had the greatest cytotoxic effect. After treatment with 30 μM octyl fenbufen amide, nearly seventy percent of the cells lost their viability. At the concentration of 10 μM, fenbufen amide analogs did not show cytotoxicity according to the MTT assay results. The NO scavenging activities of the fenbufen amide analogs were not significantly different from those of fenbufen.