MilliporeSigma
  • Development of a small scintillation detector with an optical fiber for fast neutrons.
Development of a small scintillation detector with an optical fiber for fast neutrons.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2010-12-07)
T Yagi, H Unesaki, T Misawa, C H Pyeon, S Shiroya, T Matsumoto, H Harano
ABSTRACT
To investigate the characteristics of a reactor and a neutron generator, a small scintillation detector with an optical fiber with ThO(2) has been developed to measure fast neutrons. However, experimental facilities where (232)Th can be used are limited by regulations, and S/N ratio is low because the background counts of this detector are increase by alpha decay of (232)Th. The purpose of this study is to develop a new optical fiber detector for measuring fast neutrons that does not use nuclear material such as (232)Th. From the measured and calculated results, the new optical fiber detector which uses ZnS(Ag) as a converter material together with a scintillator have the highest detection efficiency among several developed detectors. It is applied for the measurement of reaction rates generated from fast neutrons; furthermore, the absolute detection efficiency of this detector was obtained experimentally.