MilliporeSigma
  • Synthesis and functionalization of 3-alkylidene-1,2-diazetidines using transition metal catalysis.
Synthesis and functionalization of 3-alkylidene-1,2-diazetidines using transition metal catalysis.
Organic letters (2011-03-04)
Michael J Brown, Guy J Clarkson, Graham G Inglis, Michael Shipman
ABSTRACT
An efficient two-step synthesis of a wide range of 3-methylene-1,2-diazetidines has been developed through application of a Cu(I)-catalyzed 4-exo ring closure. The double bond of this new class of strained heterocycle can be functionalized in a stereocontrolled manner by using palladium-catalyzed Heck reactions. Moreover, chemoselective reduction of 3-alkylidene-1,2-diazetidines gives access to saturated 1,2-diazetidines and vicinal diamines.
Sigma-Aldrich
Azetidine, 98%