MilliporeSigma
  • Easy access to fully functionalized chiral tetrahydro-β-carboline alkaloids.
Easy access to fully functionalized chiral tetrahydro-β-carboline alkaloids.
The Journal of organic chemistry (2011-03-08)
Takayoshi Arai, Makiko Wasai, Naota Yokoyama
ABSTRACT
A four-step synthetic route to fully substituted chiral tetrahydro-β-carbolines (THBCs) is described. Starting from the (R,S,S)-Friedel-Crafts/Henry adduct obtained from three-component coupling of an indole, nitroalkene, and aldehyde catalyzed by imidazoline-aminophenol-CuOTf, the (1S,3S,4R)-THBCs were readily synthesized in a three-step operation including reduction of the nitro-functionality and Pictet-Spengler cyclization.
Sigma-Aldrich
1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole, 98%