MilliporeSigma
  • Synthesis of fluorinated purine and 1-deazapurine glycosides as potential inhibitors of adenosine deaminase.
Synthesis of fluorinated purine and 1-deazapurine glycosides as potential inhibitors of adenosine deaminase.
The Journal of organic chemistry (2011-03-15)
Viktor O Iaroshenko, Dmytro Ostrovskyi, Andranik Petrosyan, Satenik Mkrtchyan, Alexander Villinger, Peter Langer
ABSTRACT
The synthesis of 2- and 6-trifluoromethylated purines and 1-deazapurines was performed by formal [3 + 3]-cyclization reactions of 5-aminoimidazoles with a set of trifluoromethyl-substituted 1,3-CCC- and 1,3-CNC-dielectrophiles. The corresponding fluorinated nucleosides were synthesized by glycosylation of 9-unsubstituted purines and 1-deazapurines with peracetylated β-ribose, β-glucose, and rhamnose and subsequent deprotection. These scaffolds can be considered as potential inhibitors of adenosine deaminase (ADA) and inosine monophosphate dehydrogenase (IMPDH) enzymes.
Sigma-Aldrich
4-Azabenzimidazole, 99%