MilliporeSigma
  • Comparable population pharmacokinetics and pharmacodynamic breakpoints of cefpirome in cystic fibrosis patients and healthy volunteers.
Comparable population pharmacokinetics and pharmacodynamic breakpoints of cefpirome in cystic fibrosis patients and healthy volunteers.
Antimicrobial agents and chemotherapy (2011-03-16)
J B Bulitta, M Kinzig, C B Landersdorfer, U Holzgrabe, U Stephan, F Sörgel
ABSTRACT
Cystic fibrosis (CF) patients are often reported to have higher clearances and larger volumes of distribution per kilogram of total body weight (WT) for beta-lactams than healthy volunteers. As pharmacokinetic (PK) data on cefpirome from studies of CF patients are lacking, we systematically compared its population PK and pharmacodynamic breakpoints for CF patients and healthy volunteers of similar body size. Twelve adult CF patients (median lean body mass [LBM] = 45.7 kg) and 12 healthy volunteers (LBM = 50.0 kg) received a single 10-min intravenous infusion of 2 g cefpirome. Plasma and urine concentrations were determined by high-performance liquid chromatography (HPLC). Population PK and Monte Carlo simulations were performed using NONMEM and S-ADAPT and a duration of an unbound plasma concentration above the MIC ≥ 65% of the dosing interval as a pharmacodynamic target. Unscaled clearances for CF patients were similar to those seen with healthy volunteers, and the volume of distribution was 6% lower for CF patients. Linear scaling of total clearance by WT resulted in clearance that was 20% higher (P ≤ 0.001 [nonparametric bootstrap]) in CF patients. Allometric scaling by LBM explained the differences between the two subject groups with respect to average clearance and volume of distribution and reduced the unexplained between-subject variability of renal and nonrenal clearance by 10 to 14%. For the CF patients, robust (>90%) probabilities of target attainment (PTA) were achieved by the administration of a standard dose of 2 g/70 kg WT every 12 h (Q12h) given as 30-min infusions for MICs ≤ 1.5 mg/liter. As alternative dosage regimens, a 5-h infusion of 1.33 g/70 kg WT Q8h achieved robust PTAs for MICs ≤ 8 to 12 mg/liter and a continuous infusion of 4 g/day for MICs ≤ 12 mg/liter. Prolonged infusion of cefpirome is expected to be superior to short-term infusions for MICs between 2 and 12 mg/liter.