MilliporeSigma
Antibacterial properties of compounds isolated from Carpobrotus edulis.
International journal of antimicrobial agents (2011-03-18)
A Martins, A Vasas, M Viveiros, J Molnár, J Hohmann, L Amaral
ABSTRACT
Several compounds isolated from the plant Carpobrotus edulis were evaluated for their activity against multidrug-resistant (MDR) bacteria and their efflux pump systems. Amongst the compounds isolated, six compounds were tested, namely uvaol, β-amyrin, oleanolic acid, catechin, epicatechin and monogalactosyldiacylglycerol. Oleanolic acid presented high antibacterial activity against a large number of bacterial strains. The triterpene uvaol was the most active compound for modulation of efflux activity by MDR Gram-positive strains.
Sigma-Aldrich
Uvaol, ≥95%