MilliporeSigma
  • Tropisetron ameliorates ischemic brain injury in an embolic model of stroke.
Tropisetron ameliorates ischemic brain injury in an embolic model of stroke.
Brain research (2011-03-31)
Reza Rahimian, Ali Daneshmand, Shahram Ejtemaei Mehr, Anita Barzegar-Fallah, Sanaz Mohammadi-Rick, Gohar Fakhfouri, Alireza P Shabanzadeh, Ahmad Reza Dehpour
ABSTRACT
Tropisetron is widely used to counteract chemotherapy-induced emesis. Evidence obtained from human and animal studies shows that tropisetron possesses anti-inflammatory properties. In this study, we assessed the effect of tropisetron on brain damage in a rat thromboembolic model of stroke. Stroke was rendered in rats by introduction of an autologous clot into the middle cerebral artery (MCA). Tropisetron (1 or 3mg/kg); m-chlorophenylbiguanide (mCPBG), a selective 5-HT(3) receptor agonist (15 mg/kg); tropisetron (3mg/kg) plus mCPBG (15 mg/kg); granisetron (3mg/kg); tacrolimus (1mg/kg); or tacrolimus (1mg/kg) plus tropisetron (3mg/kg) were administered intraperitoneally 1h prior to embolization. Behavioral scores and infarct volume as well as myeloperoxidase (MPO) activity and tumor necrosis factor-alpha (TNF-α) level were determined in the ipsilateral cortex 4h and 48 h following stroke induction. Forty-eight hours after embolization, tropisetron (1 or 3mg/kg), tropisetron (3mg/kg) plus mCPBG (15 mg/kg), tacrolimus (1mg/kg), or tacrolimus (1mg/kg) plus tropisetron (3mg/kg) significantly curtailed brain infarction, improved behavioral scores, diminished elevated tissue MPO activity, and reduced TNF-α levels compared to control group (n=6; P<0.05). mCPBG or granisetron had no effect on the mentioned parameters. Tropisetron attenuates brain damage after a thromboembolic event. Beneficial effects of tropisetron in this setting are receptor independent.