MilliporeSigma
  • Fluindione: a francophone oral anticoagulant drug utilized in liver transplant recipients for Budd-Chiari syndrome.
Fluindione: a francophone oral anticoagulant drug utilized in liver transplant recipients for Budd-Chiari syndrome.
Journal of gastrointestinal and liver diseases : JGLD (2011-04-01)
Fabrizio Panaro, Hassan Bouyabrine, Majd Hamoui, Philippe Wolf, Francis Navarro