MilliporeSigma
Case of epirubicin-induced cardiomyopathy in familial cardiomyopathy.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2011-04-13)
Kate E Shipman, Ian Arnold