MilliporeSigma
  • Theoretical study of molecular structure and vibrational spectra of 1,4-dihydroxyanthraquinone.
Theoretical study of molecular structure and vibrational spectra of 1,4-dihydroxyanthraquinone.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-05-13)
Xiaopeng Xuan, Xinsheng Wang, Na Wang
ABSTRACT
Molecular structure and vibrational spectra of 1,4-dihydroxyanthraquinone (1,4-DAQ) are studied theoretically and experimentally. FT-infrared and Raman spectra of 1,4-DAQ are recorded in solid phase in regions of 4000-400 and 3500-100 cm(-1), respectively. The geometry of 1,4-DAQ is optimized by B3LYP and B3PW91 methods, and the most stable structure with C2v point group is found. The harmonic vibration frequencies, infrared intensities, and the Raman scattering activities of the compound are calculated, analyzed, and compared with experimental data. Our calculated frequencies agree well with the experimental results.