MilliporeSigma
  • [The transdermal 7-day buprenorphine patch--an effective and safe treatment option, if tramadol or tilidate/naloxone is insufficient. Results of a non-interventional study].
[The transdermal 7-day buprenorphine patch--an effective and safe treatment option, if tramadol or tilidate/naloxone is insufficient. Results of a non-interventional study].
MMW Fortschritte der Medizin (2011-05-20)
U Schutter, I Ritzdorf, B Heckes
ABSTRACT
The transdermal 7-day buprenorphine matrix patch provides a constant and user-friendly pain management when chronic musculoskeletal pain requires opioids. This analysis of clinical routine data evaluated the benefit of this treatment for patients previously receiving oral long-term treatment with weak opioids alone. Data of 310 patients previously treated with tramadol or tildate/naloxone and part of a multicentre observational study with 3295 patients were analyzed. In 89.7% of the 310 patients oral treatment with weak opioids was replaced by the 7-day buprenorphine patch due to insufficient analgesia. During treatment with the 7-day buprenorphine patch there was a clinically significant decrease of the mean pain intensity at rest during the day from 5.7 to 2.9, on physical effort during the day from 7.3 to 3.8 and at night from 5.2 to 2.3 (11-point NRS scale, p < or = 0.001). In addition, quality of life aspects such as mobility, self-reliance and quality of sleep improved, which are relevant for individual patient satisfaction with pain management. For patients with previous long-term tramadol or tilidate/naloxone treatment the switch to the 7-day buprenorphine matrix patch proved to be effective and safe for the management of chronic pain. The user-friendly 7-day application interval contributes to improving compliance and a reducing exposure to tablets.
Product Description
Supelco
Tilidine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®