MilliporeSigma
Ross C. Terrell, PhD, an anesthetic pioneer.
Anesthesia and analgesia (2011-06-07)
William Burns Burns, Edmond I Eger
ABSTRACT
On December 30, 2010, Ross C. Terrell, PhD, died. With his passing at age 85, we lost one of the pioneers of modern anesthesia. Terrell synthesized most of the inhalation anesthetics used today, including desflurane, enflurane, isoflurane, and sevoflurane.