MilliporeSigma
  • Total syntheses of (-) epilupinine and (-)-tashiromine using imino-aldol reactions.
Total syntheses of (-) epilupinine and (-)-tashiromine using imino-aldol reactions.
Organic letters (2011-07-13)
Amanda C Cutter, Iain R Miller, John F Keily, Richard K Bellingham, Mark E Light, Richard C D Brown
ABSTRACT
Short routes to enantiomerically pure indolizidine and quinolizidine alkaloids have been developed using imino-aldol reactions of enolates derived from phenyl 5-chlorovalerate. High levels of syn selectivity (dr ∼13-16:1) were obtained using lithium enolates of phenyl esters in combination with tert-butylsulfinyl imines. The imino-aldol adducts were deprotected and cyclized to afford (-)-epilupinine ((-)-2) and (-)-tashiromine ((-)-1) in two further steps.