MilliporeSigma
  • Synthesis of peptides containing overlapping lanthionine bridges on the solid phase: an analogue of rings D and E of the lantibiotic nisin.
Synthesis of peptides containing overlapping lanthionine bridges on the solid phase: an analogue of rings D and E of the lantibiotic nisin.
Organic letters (2011-07-20)
Begum Mothia, Antony N Appleyard, Sjoerd Wadman, Alethea B Tabor
ABSTRACT
A methodology for the solid-phase synthesis of the overlapping lanthionine bridges found in many lantibiotics has been developed. A novel Teoc/TMSE-protected lanthionine derivative has been synthesized, and this lanthionine, and an Aloc/allyl-protected lanthionine derivative, have been incorporated into a linear peptide using solid-phase peptide synthesis. Selective deprotection of the silyl protecting groups, followed by sequential cyclization, deprotection of the allyl protecting groups, and further cyclization, enabled the regioselective formation of an analogue of rings D and E of nisin.