MilliporeSigma
  • One-step isolation of γ-oryzanol from rice bran oil by non-aqueous hydrostatic countercurrent chromatography.
One-step isolation of γ-oryzanol from rice bran oil by non-aqueous hydrostatic countercurrent chromatography.
Journal of separation science (2011-07-23)
Apostolis Angelis, Aurélie Urbain, Maria Halabalaki, Nektarios Aligiannis, Alexios-Leandros Skaltsounis
ABSTRACT
The value-added γ-oryzanol was purified in one step from crude rice bran oil (RBO) using a preparative hydrostatic countercurrent chromatography (hydrostatic CCC) method, operating in the dual mode. The fractionation was performed using a non-aqueous biphasic solvent system consisting of heptane-acetonitrile-butanol (1.8:1.4:0.7, v/v/v), leading rapidly to the target compounds. Transfer of the analytical CCC method to large-scale isolation was also carried out yielding a high quantity-high purity fraction of γ-oryzanol. In addition, a fraction of hydroxylated triterpene alcohol ferulates (polar γ-oryzanol) was clearly separated and obtained. Furthermore, a fast HPLC-APCI(±)-HRMS method was developed and applied for the identification of γ-oryzanol as well as the polar γ-oryzanol in RBO and the resulting fractions. The purity of γ-oryzanol fraction was estimated as 97% based on HPLC-APCI-HRMS analysis.