MilliporeSigma
  • Reaction of O2 with [(-)-sparteine]Pd(H)Cl: evidence for an intramolecular [H-L]+ "reductive elimination" pathway.
Reaction of O2 with [(-)-sparteine]Pd(H)Cl: evidence for an intramolecular [H-L]+ "reductive elimination" pathway.
Journal of the American Chemical Society (2011-07-28)
Nattawan Decharin, Brian V Popp, Shannon S Stahl
ABSTRACT
(Sp)PdCl(2) [Sp = (-)-sparteine] catalyzes a number of different aerobic oxidation reactions, and reaction of O(2) with a Pd(II)-hydride intermediate, (Sp)Pd(H)Cl (1), is a key step in the proposed catalytic mechanism. Previous computational studies suggest that O(2) inserts into the Pd(II)-H bond, initiated by abstraction of the hydrogen atom by O(2). Experimental and computational results obtained in the present study challenge this conclusion. Oxygenation of in-situ-generated (Sp)Pd(H)Cl exhibits a zero-order dependence on [O(2)]. This result is inconsistent with a bimolecular H-atom-abstraction pathway, and DFT computational studies identify a novel "reductive elimination" mechanism, in which the chelating nitrogen ligand undergoes intramolecular deprotonation of the Pd(II)-hydride. The relevance of this mechanism to other Pd(II) oxidation catalysts with chelating nitrogen ligands is evaluated.