MilliporeSigma
  • [Secondary metabolites of seaweed endophytic fungi ZJ27 in the South China Sea coast].
[Secondary metabolites of seaweed endophytic fungi ZJ27 in the South China Sea coast].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2011-08-04)
Bi-hong Xiao, Zhi-gang She, Xiao-ling Lei, Bin Chen, Cai-huan Huang, Jia Xu
ABSTRACT
To study the secondary metabolites of coastal seaweed endophytic fungi ZJ27. The compounds were isolated by chromatographic technique. The structures were identified by comprehensive physic-chemical properties and spectral methods. Five compounds were isolated and identified as sclerotiorin (1), ochrephilone (2), 6-((3E, 5E)-5,7-dimethyl-2-methylenenona-3,5-dienyl)-2,4-dihydroxy-3-methylbenzaldehyde (3),(2E,4E)-4,6-dimethylocta-2,4-dienoic acid (4), pencolide (5). 11 g of the compound 1 and 66 mg of the compound 2 were isolated from this fungi indicate that it may develop to be medical source microorganism.
Sigma-Aldrich
m-Tolualdehyde, 97%
Pricing and availability is not currently available.