MilliporeSigma
DFT, FT-Raman and FT-IR investigations of 5-methoxysalicylic acid.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-08-16)
Esma Güneş, Cemal Parlak
ABSTRACT
FT-IR and FT-Raman spectra of 5-methoxysalicylic acid (5MeOSA) have been experimentally reported in the region of 4000-10 cm(-1) and 4000-50 cm(-1), respectively. The optimized geometric parameters, conformational equilibria, normal mode frequencies and corresponding vibrational assignments of 5MeOSA (C(8)H(8)O(4)) are theoretically examined by means of B3LYP hybrid density functional theory (DFT) method together with 6-31++G(d,p) basis set. Furthermore, reliable vibrational assignments have made on the basis of potential energy distribution (PED) calculated and the thermodynamics functions, highest occupied and lowest unoccupied molecular orbitals (HOMO and LUMO) of 5MeOSA have been predicted. Calculations are employed for different conformations of 5MeOSA, both in gas phase and in solution. Solvent effects are investigated using chloroform and dimethylsulfoxide. All results indicate that B3LYP method is able to provide satisfactory results for predicting vibrational frequencies and the structural parameters, vibrational frequencies and assignments, IR and Raman intensities of 5MeOSA are solvent dependent.
Sigma-Aldrich
2-Hydroxy-5-methoxybenzoic acid, 98%