MilliporeSigma
  • The extraction of aromatic carboxylic acids by the copper complex with Curtis macrocyclic tetramine and its utilization for photometric determination of nonsteroidal anti-inflammatory drugs.
The extraction of aromatic carboxylic acids by the copper complex with Curtis macrocyclic tetramine and its utilization for photometric determination of nonsteroidal anti-inflammatory drugs.
Acta poloniae pharmaceutica (2011-09-21)
Ivan Zseltvay, Olga Zheltvay, Valerij Antonovich
ABSTRACT
Copper complex with Curtis macrocyclic tetramine is offered as reagent for extraction-photometric determination of nonsteroidal anti-inflammatory drugs (NSAIDs), belonging to the class of aromatic carboxylic acids. The studies indicate that this method is suitable for quantitative determination of NSAIDs, which have the constant distribution in the system chloroform/water (log P) no less than 3 and dissolubility in chloroform (S) no less than 10 mg/mL. Under optimum conditions, there are liner relationships between the absorption of chloroform extracts and concentration of NSAID in the range of 0.2-4 mg/mL for indometacin (Ind), 0.2-3 mg/mL for mefenamic acid (Mef) and 0.5-3 mg/mL for diclofenac (Dic). The detection limits (S/N = 3) of Ind, Mef and Dic are 0.2, 0.1 and 0.15 mg/mL, respectively. With the help of calculating method (SPARC V4.2) it was predicted the possibility of utilization of this method for extractive-photometric determination of its detached specimen NSAID.
Product Description
Supelco
Tetramethylenedisulfotetramine solution, 100 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®