MilliporeSigma
  • [Efficacy of cortexin in the treatment of memory disorders in children].
[Efficacy of cortexin in the treatment of memory disorders in children].
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova (2011-10-27)
L S Chutko, S Iu Surushkina, E A Iakovenko, Iu L Bykova, I S Nikishena
ABSTRACT
Children, aged from 7 to 12 years, with memory disorders were treated with cortexin (30 patients) and encephabol (30 patients). The comparative evaluation of efficacy and safety of these drugs was carried out. The higher cortexin efficacy (the improvement in 86.7% of cases) in comparison with encephabol (the improvement in 63.3% of cases) confirmed by the data of neuropsychological and neurophysiological research is established.