MilliporeSigma
Efficient improved extraction of tomato saponin using shock waves.
Chemical & pharmaceutical bulletin (2011-11-02)
Hideyuki Manabe, Ayumi Takemoto, Hiromichi Maehara, Mizuho Ohno, Yoshihiro Murakami, Shigeru Itoh, Toshihiro Nohara
ABSTRACT
In the conventional method of mixer blending extraction, the yields of the tomato-saponin, esculeoside A, in the mini and middy tomatoes were found to be 0.043% and 0.046%, respectively. In order to improve the yields, we attempted a more efficient extraction using shock waves. The yields of esculeoside A were 0.0987% in air after 1 shock, 0.0792% in air after two shots, 0.0648% in half water after 1 or 2 shocks, and 0.0599% in water after 1 or 2 shocks. The yields obtained by the proposed method were approximately twice those of the conventional mixer blending method; therefore, this method is regarded to be very efficient. Moreover, two spirosolane glycosides, tomatine and lycoperoside A, were obtained for the first time from the ripe tomato fruit in this method. To date, these compounds have not been obtained with the mixer blending method. However, whether these glycosides are produced from esculeoside A or are newly extracted from the plant organ by the shock wave is still unclear.