MilliporeSigma
  • A simple synthesis of APM ([p-(N-acrylamino)-phenyl]mercuric chloride), a useful tool for the analysis of thiolated biomolecules.
A simple synthesis of APM ([p-(N-acrylamino)-phenyl]mercuric chloride), a useful tool for the analysis of thiolated biomolecules.
Bioorganic & medicinal chemistry letters (2011-11-15)
V Lemau de Talancé, F Bauer, D Hermand, S P Vincent
ABSTRACT
This study describes two novel synthetic procedures to prepare APM, a useful tool for the analysis and the purification of thiolated biomolecules. The methods developed are technically simple and robust and allowed the first full characterization of pure APM. Moreover, the efficacy of APM, as a biochemical tool, was demonstrated by analysis of tRNA thiolation by APM-PAGE.