MilliporeSigma
  • Signalling and chemosensitivity assays in melanoma: is mutated status a prerequisite for targeted therapy?
Signalling and chemosensitivity assays in melanoma: is mutated status a prerequisite for targeted therapy?
Experimental dermatology (2011-11-19)
Thierry Passeron, Jean-Philippe Lacour, Maryline Allegra, Coralie Ségalen, Anne Deville, Antoine Thyss, Damien Giacchero, Jean-Paul Ortonne, Corine Bertolotto, Robert Ballotti, Philippe Bahadoran
ABSTRACT
Selection for targeted therapies in melanoma is currently based on the search for mutations in selected genes. We aimed at evaluating the interest of signalling and chemosensitivity studies in addition to genotyping for assessing the best suitable treatment in an individual patient. We extracted genomic DNA and melanoma cells from tumor tissue of a skin metastasis of a 17-year-old woman with stage IV melanoma progressing despite three successive lines of treatment. Despite the absence of mutation in BRAF, NRAS cKIT, the MAPK pathway was activated and a significant response to sorafenib, a mitogen-activated protein kinase (MAPK)/RAF inhibitor, was found in signalling and chemosensitivity assays. A treatment combining sorafenib and dacarbazine produced a partial response for 9 months, with marked necrosis in some lesions. Chemosensitivity assays and signalling pathway studies could be of great value in addition to genotyping for assessing the most appropriate treatment in melanoma.