MilliporeSigma
  • Expanding the azaspiro[3.3]heptane family: synthesis of novel highly functionalized building blocks.
Expanding the azaspiro[3.3]heptane family: synthesis of novel highly functionalized building blocks.
Organic letters (2011-11-25)
Johannes A Burkhard, Carine Guérot, Henner Knust, Erick M Carreira
ABSTRACT
The preparation of versatile azaspiro[3.3]heptanes carrying multiple exit vectors is disclosed. Expedient synthetic routes enable the straightforward access to these novel modules that are expected to have significance in drug discovery and design.
Sigma-Aldrich
Trimethylene oxide, 97%