MilliporeSigma
  • Assessment of the health risks related to the presence of drug residues in water for human consumption: application to carbamazepine.
Assessment of the health risks related to the presence of drug residues in water for human consumption: application to carbamazepine.
Regulatory toxicology and pharmacology : RTP (2011-12-20)
Paul Houeto, Aude Carton, Michel Guerbet, Anne-Cécile Mauclaire, Chantal Gatignol, Philippe Lechat, Dominique Masset
ABSTRACT
Pharmaceutical residues have been detected at low (usually ng/L) concentrations in drinking water sources. The detection of drugs in water intended for human consumption (WIHC) has raised questions of safety. In the absence of regulatory or other official guidance, water utilities are faced with a problem of which pharmaceutical residues should be monitored and the toxicological limits that should be required. In this essay, we define an approach for the assessment of health risks related to chemicals found in drinking water. We use the examples of carbamazepine and its main metabolite 10,11-epoxycarbamazepine to demonstrate our approach, which involves application of the following algorithm: (1) when there is human or animal toxicity data, a toxicity reference value (TRV) can be calculated; (2) when this is not applicable, an attempt should be made to derive the TRV using known information about the minimum therapeutic dose (MTD); and (3) when no applicable data is available, at all, a threshold of toxicological concern (TTC) should be estimated. In the case of carbamazepine, where relevant toxicological data exists, we derived a TRV, based on the known minimum therapeutic dose (MTD). For carbamazepine's metabolite 10,11-epoxycarbamazepine, there is no toxicological data, so we applied the TTC approach. Using this approach, and combining our estimates with what is known about these chemicals' margin of exposure (MOE), suggests that there is likely to be no appreciable risk to human health exposure to carbamazepine or its major metabolite, even given the inevitable uncertainties in exposure scenarios.
Product Description
Supelco
Carbamazepine-10,11-epoxide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Carbamazepine 10,11-epoxide, analytical standard