MilliporeSigma
  • Bufothionine, a possible effective component in cinobufocini injection for hepatocellular carcinoma.
Bufothionine, a possible effective component in cinobufocini injection for hepatocellular carcinoma.
Journal of ethnopharmacology (2012-01-03)
Rui-Fang Xie, Zhi-Cheng Li, Bo Gao, Zhi-Na Shi, Xin Zhou
ABSTRACT
Cinobufacini has been traditionally used in China for the treatment of tumor since hundreds years ago. For recent years, its modern preparation,cinobifucini injection has also obtained satisfactory therapeutic functions for cancer. High performance liquid chromatography (HPLC) analysis was applied to determine the content of cinobufagin, resibufogenin and bufothionine in cinobufacin extract liquid and injection; MTT assay and flow cytometric analysis were also respectively used to study the effect of cinobufacini extract liquid, injection and three chemical structures on cells and cell cycles. HPLC results demonstrated that in cinobufacini extract liquid three ingredients (cinobufagin, resibufogenin and bufothionine) were all monitored while in cinofacini injection only bufothinone was detected; MTT assays showed bufothionine could obviously inhibit the proliferation of human hepatocellular carcinoma cell lines such as SMMC-7721 and BEL-7402 in a dose- and time-dependent manner as well as cinobufagin and resibufogenin; further flow cytometric analysis indicated obvious increases in G2/M phase and decrease in G0/G1 phase when SMMC-7721 cell line exposure to bufothionine (480 μg/ml). These results suggested bufothionine could be involved in treatment of human cancer for cinobufacini injection and the mechanism might be relative to induce G2/M phase cell cycle arrest.