MilliporeSigma
Rice bran lysolecithin as a source of energy in broiler chicken diet.
British poultry science (2012-01-10)
M V L N Raju, S V Rama Rao, P P Chakrabarti, B V S K Rao, A K Panda, B L A Prabhavathi Devi, V Sujatha, J R C Reddy, G Shyam Sunder, R B N Prasad
ABSTRACT
1. Rice bran lysolecithin (RBL) was evaluated in broiler chicken diets. In the first experiment, RBL was included in diet at 0, 0·5, 2, 8 and 32 g/kg and fed to 250 broiler chickens from 0 to 42 d of age. In the second experiment, RBL was fed at 0, 25 and 50 g/kg diet to 405 day-old broiler chickens until 21 d of age, while during the finisher phase (22-35 d of age) chickens receiving each concentration of RBL were given all three concentrations of RBL in a 3 × 3 factorial manner. The diets were isocaloric. 2. Body weight, food consumption and food conversion efficiency were unaffected by feeding RBL, while the weight of pancreas increased at ≥2 g/kg of RBL in diet (experiment 1). In experiment 2, body weight was greater in the chickens receiving RBL at either 25 or 50 g/kg (21 d) and 50 g/kg (35 d of age). At 21 d of age, food consumption was greater at 25 or 50 g RBL/kg diet, while food conversion efficiency improved with 50 g RBL/kg diet. 3. Fat digestibility increased with RBL at 32 g/kg (experiment 1) and ≤25 g/kg (experiment 2). Rice bran lysolecithin increased ready to cook weight at 50 g/kg during starter phase and decreased abdominal fat at 25 and 50 g/kg during finisher phase (experiment 2). Liver and meat fat content were not affected. 4. It is concluded that lysolecithin from rice bran oil could be used as energy supplement in broiler chicken diet.