MilliporeSigma
Intercalating oleylamines in graphite oxide.
Langmuir : the ACS journal of surfaces and colloids (2012-01-11)
Kaikun Yang, Si Liang, Lianfeng Zou, Liwei Huang, Cheol Park, Lisheng Zhu, Jiye Fang, Qiang Fu, Howard Wang
ABSTRACT
Graphite oxide has been synthesized from raw graphite particles and been treated with various mass amounts of oleylamine as intercalants to form intercalation compounds. X-ray diffraction patterns reveal that the inter-sheet distances strongly depend on the graphite oxide to oleylamine mass ratios. The equilibrium-like behavior implies diffusion-dominated oleylamine adsorption on graphite oxide in solution and excluded volume intercalations among oleylamine-adsorbed graphite oxide during restacking. The intercalation compounds are soluble in organic solvents, and their applications in the fabrication of transparent and conductive coatings have been demonstrated.
Sigma-Aldrich
Oleylamine, technical grade, 70%