MilliporeSigma
  • Effects of graded levels of dietary methionine hydroxy analogue on immune response and antioxidant status of immune organs in juvenile Jian carp (Cyprinus carpio var. Jian).
Effects of graded levels of dietary methionine hydroxy analogue on immune response and antioxidant status of immune organs in juvenile Jian carp (Cyprinus carpio var. Jian).
Fish & shellfish immunology (2012-01-17)
Sheng-Yao Kuang, Wei-Wei Xiao, Lin Feng, Yang Liu, Jun Jiang, Wei-Dan Jiang, Kai Hu, Shu-Hong Li, Ling Tang, Xiao-Qiu Zhou
ABSTRACT
Immune response and antioxidant status of immune organs in juvenile Jian carp (Cyprinus carpio var. Jian) fed graded levels of methionine hydroxy analogue (MHA) (0, 5.1, 7.6, 10.2, 12.7, 15.3 g kg(-1) diet) for 60 days were investigated. Results indicated that head kidney index, spleen index, red and white blood cell counts significantly increased with increasing MHA levels up to a point (P < 0.05), whereupon decreased (P < 0.05). Glutathione reductase activity in head kidney and spleen, anti-hydroxy radical and glutathione-S-transferase activities in spleen, catalase activity and GSH content in head kidney significantly increased by MHA supplement, while malondialdehyde content, anti-superoxide anion, superoxide dismutase, glutathione peroxidase activities in head kidney and spleen, protein carbonyl content and catalase activity in spleen, anti-hydroxy radical activity in head kidney significantly decreased by MHA supplement. However, protein carbonyl content and glutathione-S-transferase activity in head kidney, GSH content in spleen remained unaffected. After 60-day feeding trial, a challenge study was conducted by injection of Aeromonas hydrophila for 17 days. Results showed that survival rate, leukocytes phagocytic activity, lysozyme activity, acid phosphatase activity, total iron-binding capacity, haemagglutination titre, complement 3, 4 and immunoglobulin M contents significantly increased by optimal dietary MHA supplement (P < 0.05). These data suggested that MHA affected antioxidant status of immune organs and promoted immune response in juvenile Jian carp.
Sigma-Aldrich
2-Hydroxy-4-(methylthio)butyric acid calcium salt, ≥99.0% (NT)
Pricing and availability is not currently available.