MilliporeSigma
  • Genotoxicity of 7H-dibenzo[c,g]carbazole and its methyl derivatives in human keratinocytes.
Genotoxicity of 7H-dibenzo[c,g]carbazole and its methyl derivatives in human keratinocytes.
Mutation research (2012-02-07)
Zuzana Valovičová, Monika Mesárošová, Lenka Trilecová, Eva Hrubá, Soňa Marvanová, Pavel Krčmář, Alena Milcová, Jana Schmuczerová, Jan Vondráček, Miroslav Machala, Jan Topinka, Alena Gábelová
ABSTRACT
Differences between tissues in the expression of drug-metabolizing enzymes may substantially contribute to tissue-specificity of chemical carcinogens. To verify this hypothesis, the spontaneously immortalized human keratinocytes HaCaT were used, in order to evaluate the genotoxic potential of 7H-dibenzo[c,g]carbazole (DBC), a known hepatocarcinogen and sarcomagen, and its synthetic tissue-specific derivatives, 5,9-dimethyl-DBC (DiMeDBC) and N-methyl-DBC (N-MeDBC), which manifest specific tropism to the liver and skin, respectively. HaCaT cells mainly express cytochrome P4501A1 (CYP1A1), which is involved in metabolism of DBC and N-MeDBC, but not DiMeDBC [10]. Both DBC and the sarcomagen N-MeDBC induced significant levels of DNA strand-breaks, micronuclei, and DNA adducts followed by the phosphorylation of the p53 protein and histone H2AX in HaCaT cells. In contrast, the specific hepatocarcinogen DiMeDBC was devoid of any significant genotoxic activity in this cell line. Our study demonstrates that the absence of drug-metabolizing enzyme(s) involved in DiMeDBC metabolism may contribute substantially to the tissue-specific genotoxicity of this hepatocarcinogen.
Product Description
7H-Dibenzo[c,g]carbazole, BCR®, certified reference material